4-Chlor-6,7-dimethoxychinolin 98%

BLDPBD12928-100GEA 278 EUR
BLDPBD12928-100G BLDPBD12928-5G BLDPBD12928-1G BLDPBD12928-10G BLDPBD12928-25G
4-Chlor-6,7-dimethoxychinolin 98%
4-Chloro-6,7-dimethoxyquinoline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₁H₁₀ClNO₂
Molekulargewicht: 223,6556 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07778437
CAS-Nummer: 35654-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR