Ethyl-[4-(trifluormethyl)phenyl]acetat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD12997-100GEA 608 EUR
BLDPBD12997-100G BLDPBD12997-10G BLDPBD12997-250MG BLDPBD12997-5G BLDPBD12997-1G BLDPBD12997-25G
Ethyl-[4-(trifluormethyl)phenyl]acetat 98%
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₁F₃O₂
Molekulargewicht: 232,20 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07779351
CAS-Nummer: 721-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR