8-Methoxychinaldin 95%

BLDPBD13024-100MGEA 15 EUR
BLDPBD13024-100MG BLDPBD13024-250MG BLDPBD13024-1G
8-Methoxychinaldin 95%
8-Methoxychinaldin

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₁NO
Molekulargewicht: 173,2111 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00034436
CAS-Nummer: 3033-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR