Forskolin 98%

BLDPBD131541-250MGEA 76 EUR
BLDPBD131541-250MG BLDPBD131541-1G BLDPBD131541-100MG BLDPBD131541-5G
Forskolin 98%
Forskolin

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₂H₃₄O₇
Molekulargewicht: 410,51 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00082317
CAS-Nummer: 66575-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR