3-Morpholinobenzaldehyd 98%

BLDPBD13194-100MGEA 30 EUR
BLDPBD13194-100MG BLDPBD13194-5G BLDPBD13194-1G BLDPBD13194-250MG
3-Morpholinobenzaldehyd 98%
3-Morpholinobenzaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₃NO₂
Molekulargewicht: 191,2264 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07772811
CAS-Nummer: 446866-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR