Geranylacetat 70%

BLDPBD131966-100GEA 19 EUR
BLDPBD131966-100G BLDPBD131966-500G BLDPBD131966-0.5G BLDPBD131966-1000G BLDPBD131966-25G
Geranylacetat 70%
Geranylacetat

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₂₀O₂
Molekulargewicht: 196,29 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00015037
CAS-Nummer: 105-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR