4-(4-Morpholinylcarbonyl)phenylboronsäure 98%

BLDPBD13217-250MGEA 23 EUR
BLDPBD13217-250MG BLDPBD13217-1G BLDPBD13217-5G
4-(4-Morpholinylcarbonyl)phenylboronsäure 98%
4-(4-Morpholinylcarbonyl)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₄BNO₄
Molekulargewicht: 235,05 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03411952
CAS-Nummer: 389621-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR