4-Pentylphenylboronsäure 97%
|
|
BLDPBD13408-10GEA
21
EUR
BLDPBD13408-10G
BLDPBD13408-25G
BLDPBD13408-5G
BLDPBD13408-1G
BLDPBD13408-100G
4-Pentylphenylboronsäure 97%
4-Pentylphenylboronsäure
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₁₁H₁₇BO₂ Molekulargewicht: 192,07 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00995151 CAS-Nummer: 121219-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...