Indium(III)chlorid Tetrahydrat 98+%

BLDPBD134586-25GEA 42 EUR
BLDPBD134586-25G BLDPBD134586-5G
Indium(III)chlorid Tetrahydrat 98+%
Indium(III)chlorid Tetrahydrat

Storage condition: Inert atmosphere, Room Temperature

Formel: Cl₃In·4H₂O
Molekulargewicht: 293,238 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149746
CAS-Nummer: 22519-64-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR