4-Amino-1-Boc-piperidin-4-carbonsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD13474-5GEA 73 EUR
BLDPBD13474-5G BLDPBD13474-1G BLDPBD13474-10G BLDPBD13474-250MG
4-Amino-1-Boc-piperidin-4-carbonsäure 97%
4-Amino-1-Boc-piperidin-4-carbonsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₁H₂₀N₂O₄
Molekulargewicht: 244,29 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01318728
CAS-Nummer: 183673-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR