(±)-Isoborneol 90%
|
|
BLDPBD135004-5GEA
7
EUR
BLDPBD135004-5G
BLDPBD135004-25G
(±)-Isoborneol 90%
(±)-Isoborneol
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₀H₁₈O Molekulargewicht: 154,2493 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00074821 CAS-Nummer: 124-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...