2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester 98%

BLDPBD1366-100MGEA 80 EUR
BLDPBD1366-100MG BLDPBD1366-250MG BLDPBD1366-1G
2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester 98%
2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₁H₂₂BNO₄
Molekulargewicht: 363,215 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02179492
CAS-Nummer: 380430-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR