Methyl-2-methoxyisonicotinat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD138325-5GEA 199 EUR
BLDPBD138325-5G BLDPBD138325-250MG BLDPBD138325-1G
Methyl-2-methoxyisonicotinat 98%
Methyl-2-methoxyisonicotinat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₉NO₃
Molekulargewicht: 167,1620 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08436071
CAS-Nummer: 26156-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR