Methyl-3,5-bis(trifluormethyl)benzoat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD138501-10GEA 19 EUR
BLDPBD138501-10G BLDPBD138501-100G BLDPBD138501-5G BLDPBD138501-1G BLDPBD138501-25G
Methyl-3,5-bis(trifluormethyl)benzoat 98%
Methyl-3,5-bis(trifluormethyl)benzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₆F₆O₂
Molekulargewicht: 272,15 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00052317
CAS-Nummer: 26107-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR