Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat 98%
|
|
BLDPBD138617-250MGEA
25
EUR
BLDPBD138617-250MG
BLDPBD138617-1G
BLDPBD138617-100MG
BLDPBD138617-5G
Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat 98%
Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Storage condition: Sealed in dry, 2-8°C
Formel:
C₉H₇BrN₂O₂ Molekulargewicht: 255,0681 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07021501 CAS-Nummer: 886361-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...