Methyl-4-amino-3-nitrobenzoat 95%
|
|
BLDPBD138892-250MGEA
6
EUR
BLDPBD138892-250MG
BLDPBD138892-100G
BLDPBD138892-5G
BLDPBD138892-25G
BLDPBD138892-1G
BLDPBD138892-10G
Methyl-4-amino-3-nitrobenzoat 95%
Methyl-4-amino-3-nitrobenzoat
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₈H₈N₂O₄ Molekulargewicht: 196,16 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00017562 CAS-Nummer: 3987-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...