myo-Inosit 98%

Synonyme: cis-1,2,3,5-trans-4,6-Cyclohexanhexolmeso-Inositolmyo-Inositolmeso-Inosit

BLDPBD140573-5GEA 6 EUR
BLDPBD140573-5G BLDPBD140573-0.025 BLDPBD140573-1000G BLDPBD140573-100G BLDPBD140573-500G BLDPBD140573-25G
myo-Inosit 98%
myo-Inosit

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₁₂O₆
Molekulargewicht: 180,16 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00077932
CAS-Nummer: 87-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR