Bis(pinakolato)dibor 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD14304-500GEA 136 EUR
BLDPBD14304-500G BLDPBD14304-25G BLDPBD14304-10G BLDPBD14304-100G BLDPBD14304-5G BLDPBD14304-1000G BLDPBD14304-0.5G
Bis(pinakolato)dibor 98%
Bis(pinakolato)dibor

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₂₄B₂O₄
Molekulargewicht: 253,94 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00799570
CAS-Nummer: 73183-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR