N-Benzyl-4-brombenzolsulfonamid 98%

BLDPBD143422-25GEA 170 EUR
BLDPBD143422-25G BLDPBD143422-5G BLDPBD143422-1G
N-Benzyl-4-brombenzolsulfonamid 98%
N-Benzyl-4-brombenzolsulfonamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₂BrNO₂S
Molekulargewicht: 326,21 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00450587
CAS-Nummer: 3609-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR