2-Pyrid-3-ylbenzoesäure 98%

BLDPBD14467-100MGEA 22 EUR
BLDPBD14467-100MG BLDPBD14467-5G BLDPBD14467-250MG BLDPBD14467-1G
2-Pyrid-3-ylbenzoesäure 98%
2-Pyrid-3-ylbenzoesäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₉NO₂
Molekulargewicht: 199,2054 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03426511
CAS-Nummer: 134363-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR