4-Pyrid-2-ylbenzoesäure 98%

BLDPBD14473-5GEA 300 EUR
BLDPBD14473-5G BLDPBD14473-250MG BLDPBD14473-1G
4-Pyrid-2-ylbenzoesäure 98%
4-Pyrid-2-ylbenzoesäure

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₉NO₂
Molekulargewicht: 199,2100 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01830093
CAS-Nummer: 4385-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR