2-Amino-4'-brombenzophenon 98%

BLDPBD14495-100GEA 467 EUR
BLDPBD14495-100G BLDPBD14495-1G BLDPBD14495-5G BLDPBD14495-25G
2-Amino-4'-brombenzophenon 98%
2-Amino-4'-brombenzophenon

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₃H₁₀BrNO
Molekulargewicht: 276,1300 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02181034
CAS-Nummer: 1140-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR