4-(Benzyloxy)phenylboronsäure 98%

BLDPBD14647-5GEA 11 EUR
BLDPBD14647-5G BLDPBD14647-25G BLDPBD14647-1G BLDPBD14647-10G BLDPBD14647-100G
4-(Benzyloxy)phenylboronsäure 98%
4-(Benzyloxy)phenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₃H₁₃BO₃
Molekulargewicht: 228,06 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01075705
CAS-Nummer: 146631-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR