Kalium(2-(methylsulfonyl)phenyl)trifluoroborat 98%

BLDPBD148653-1GEA 84 EUR
BLDPBD148653-1G BLDPBD148653-5G BLDPBD148653-250MG
Kalium(2-(methylsulfonyl)phenyl)trifluoroborat 98%
Potassium-2-(methylsulphonyl)phenyltrifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₇BF₃KO₂S
Molekulargewicht: 262,1000 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04115760
CAS-Nummer: 850623-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR