4-Carbamoylphenylboronsäurepinakolester 95%
Lieferant: BLD PHARMATECH GMBH
BLDPBD14888-1GEA
24
EUR
BLDPBD14888-1G
BLDPBD14888-250MG
BLDPBD14888-5G
4-Carbamoylphenylboronsäurepinakolester 95%
4-Carbamoylphenylboronsäurepinakolester
Formel:
C₁₃H₁₈BNO₃ Molekulargewicht: 247,0979 g/mol |
MDL Number:
MFCD05663887 CAS-Nummer: 179117-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...