(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid 97%

Synonyme: (2S)-2-(3,4,5-Trimethoxyphenyl)butyric acid(S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid

BLDPBD14936-1GEA 130 EUR
BLDPBD14936-1G BLDPBD14936-25G BLDPBD14936-100MG BLDPBD14936-5G BLDPBD14936-10G BLDPBD14936-250MG
(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid 97%
(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₈O₅
Molekulargewicht: 254,279 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06245370
CAS-Nummer: 195202-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR