Purin 98%
|
|
Synonyme:
7H-Imidazo(4,5-d)pyrimidin
BLDPBD149554-25GEA
576
EUR
BLDPBD149554-25G
BLDPBD149554-250MG
BLDPBD149554-5G
BLDPBD149554-10G
BLDPBD149554-1G
Purin 98%
Purin
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₅H₄N₄ Molekulargewicht: 120,11 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00079221 CAS-Nummer: 120-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...