4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid 97%

BLDPBD149638-250MGEA 66 EUR
BLDPBD149638-250MG BLDPBD149638-5G BLDPBD149638-100MG BLDPBD149638-1G
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid 97%
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₁₀N₂O₂
Molekulargewicht: 166,177 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09056814
CAS-Nummer: 307313-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR