5-Brom-4-methylpyrimidin 98%

BLDPBD149845-10GEA 338 EUR
BLDPBD149845-10G BLDPBD149845-25G BLDPBD149845-5G BLDPBD149845-1G BLDPBD149845-250MG
5-Brom-4-methylpyrimidin 98%
5-Bromo-4-methylpyrimidine

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₅BrN₂
Molekulargewicht: 173,0100 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10000597
CAS-Nummer: 1439-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR