2-Amino-2'-chlor-5-nitrobenzophenon 97%
|
|
BLDPBD15101-25GEA
42
EUR
BLDPBD15101-25G
BLDPBD15101-10G
BLDPBD15101-5G
BLDPBD15101-100G
2-Amino-2'-chlor-5-nitrobenzophenon 97%
2-Amino-2'-chlor-5-nitrobenzophenon
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₁₃H₉ClN₂O₃ Molekulargewicht: 276,68 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00792453 CAS-Nummer: 2011-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...