Spermidin Trihydrochlorid 99+%

Synonyme: 4-Azaoctamethylendiamin TrihydrochloridN-(3-Aminopropyl)-1,4-diaminobutan Trihydrochlorid4-Azoniaoctamethylendiammoniumtrichlorid

BLDPBD151797-1GEA 49 EUR
BLDPBD151797-1G BLDPBD151797-250MG BLDPBD151797-25G BLDPBD151797-5G
Spermidin Trihydrochlorid 99+%
Spermidin Trihydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₂₂Cl₃N₃
Molekulargewicht: 254,63 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012918
CAS-Nummer: 334-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR