1-(Phenylsulfonyl)-3-indolboronsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD15240-1GEA 93 EUR
BLDPBD15240-1G BLDPBD15240-250MG BLDPBD15240-10G BLDPBD15240-5G BLDPBD15240-25G
1-(Phenylsulfonyl)-3-indolboronsäure 95%
1-(Phenylsulphonyl)-3-indoleboronic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₄H₁₂BNO₄S
Molekulargewicht: 301,1300 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02681892
CAS-Nummer: 129271-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR