5-Indolboronsäurepinakolester 97%

BLDPBD15494-100MGEA 7 EUR
BLDPBD15494-100MG BLDPBD15494-1G BLDPBD15494-10G BLDPBD15494-100G BLDPBD15494-25G BLDPBD15494-250MG BLDPBD15494-5G
5-Indolboronsäurepinakolester 97%
5-Indolboronsäurepinakolester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₄H₁₈BNO₂
Molekulargewicht: 243,1092 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03789263
CAS-Nummer: 269410-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR