Tris[3,5-bis(trifluormethyl)phenyl]phosphin 95%

BLDPBD155935-1GEA 89 EUR
BLDPBD155935-1G BLDPBD155935-250MG BLDPBD155935-100MG BLDPBD155935-5G
Tris[3,5-bis(trifluormethyl)phenyl]phosphin 95%
Tris[3,5-bis(trifluormethyl)phenyl]phosphin

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₄H₉F₁₈P
Molekulargewicht: 670,28 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00042428
CAS-Nummer: 175136-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR