3-Chloro-2-nitroanisole 96%

BLDPBD158243-1GEA 79 EUR
BLDPBD158243-1G BLDPBD158243-250MG BLDPBD158243-5G
3-Chloro-2-nitroanisole 96%
3-Chloro-2-nitroanisole

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₆ClNO₃
Molekulargewicht: 187,58 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD14455916
CAS-Nummer: 5472-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR