4-Brom-2-fluor-6-nitroanilin 98%

BLDPBD158353-10GEA 70 EUR
BLDPBD158353-10G BLDPBD158353-100G BLDPBD158353-25G BLDPBD158353-5G BLDPBD158353-250MG BLDPBD158353-1G
4-Brom-2-fluor-6-nitroanilin 98%
4-Brom-2-fluor-6-nitroanilin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₄BrFN₂O₂
Molekulargewicht: 235,0100 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03094271
CAS-Nummer: 517920-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR