4-Chlor-3-(cyclopropylcarbamoyl)phenylboronsäure 98%

BLDPBD158595-250MGEA 66 EUR
BLDPBD158595-250MG BLDPBD158595-100MG BLDPBD158595-1G
4-Chlor-3-(cyclopropylcarbamoyl)phenylboronsäure 98%
4-Chlor-3-(cyclopropylcarbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₁BClNO₃
Molekulargewicht: 239,46 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07783865
CAS-Nummer: 871332-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR