5-Brom-2-methyl-2H-indazol 97%

BLDPBD158608-250MGEA 7 EUR
BLDPBD158608-250MG BLDPBD158608-100G BLDPBD158608-5G BLDPBD158608-100MG BLDPBD158608-10G BLDPBD158608-1G BLDPBD158608-25G
5-Brom-2-methyl-2H-indazol 97%
5-Bromo-2-methyl-2H-indazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇BrN₂
Molekulargewicht: 211,0600 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09870047
CAS-Nummer: 465529-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR