5-Bromo-1H-imidazo[4,5-b]pyridine, 95%
|
|
BLDPBD159126-250MGEA
37
EUR
BLDPBD159126-250MG
5-Bromo-1H-imidazo[4,5-b]pyridine, 95%
5-Bromo-1H-imidazo[4,5-b]pyridine,
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₆H₄BrN₃ Molekulargewicht: 198,02 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD11227145 CAS-Nummer: 28279-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...