Methyl-2-amino-5-bromisonicotinat 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD159364-25GEA
394
EUR
BLDPBD159364-25G
BLDPBD159364-10G
BLDPBD159364-100G
BLDPBD159364-1G
BLDPBD159364-250MG
BLDPBD159364-5G
BLDPBD159364-0.025
Methyl-2-amino-5-bromisonicotinat 98%
Methyl-2-amino-5-bromisonicotinat
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₇H₇BrN₂O₂ Molekulargewicht: 231,0467 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09027294 CAS-Nummer: 882499-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...