2-Chloro-5-(trifluoromethyl)nicotinic acid 98%

BLDPBD159700-250MGEA 9 EUR
BLDPBD159700-250MG BLDPBD159700-5G BLDPBD159700-10G BLDPBD159700-1G BLDPBD159700-100MG BLDPBD159700-100G BLDPBD159700-25G
2-Chloro-5-(trifluoromethyl)nicotinic acid 98%
2-Chloro-5-(trifluoromethyl)nicotinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₃ClF₃NO₂
Molekulargewicht: 225,55 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08741353
CAS-Nummer: 505084-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR