2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane 97%

BLDPBD159797-250MGEA 38 EUR
BLDPBD159797-250MG BLDPBD159797-100MG BLDPBD159797-1G
2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane 97%
2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₈BClO₃
Molekulargewicht: 268,544 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07782041
CAS-Nummer: 1151564-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR