6-Bromoquinoline-3-carboxylic acid 97%

BLDPBD160157-10GEA 765 EUR
BLDPBD160157-10G BLDPBD160157-250MG BLDPBD160157-5G BLDPBD160157-1G BLDPBD160157-25G
6-Bromoquinoline-3-carboxylic acid 97%
6-Bromoquinoline-3-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₆BrNO₂
Molekulargewicht: 252,064 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08437553
CAS-Nummer: 798545-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR