(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD160440-25GEA 382 EUR
BLDPBD160440-25G BLDPBD160440-1G BLDPBD160440-5G BLDPBD160440-250MG BLDPBD160440-10G
(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate 97%
(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₇NO₄S
Molekulargewicht: 259,32 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00040622
CAS-Nummer: 111769-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR