3-Fluor-4-(trifluormethoxy)anilin 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD160892-5GEA 104 EUR
BLDPBD160892-5G BLDPBD160892-100MG BLDPBD160892-1G BLDPBD160892-25G BLDPBD160892-250MG BLDPBD160892-10G BLDPBD160892-100G
3-Fluor-4-(trifluormethoxy)anilin 97%
3-Fluor-4-(trifluormethoxy)anilin

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₅F₄NO
Molekulargewicht: 195,12 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09832383
CAS-Nummer: 1017779-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR