2-Methyl-4'-(methylthio)-2-morpholinopropiophenon 98%

BLDPBD16092-25GEA 7 EUR
BLDPBD16092-25G BLDPBD16092-10G
2-Methyl-4'-(methylthio)-2-morpholinopropiophenon 98%
2-Methyl-4'-(methylthio)-2-morpholinopropiophenon

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₅H₂₁NO₂S
Molekulargewicht: 279,3977 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00083014
CAS-Nummer: 71868-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR