2-Chlor-6-hydroxybenzonitril 97%

BLDPBD161281-25GEA 192 EUR
BLDPBD161281-25G BLDPBD161281-5G BLDPBD161281-1G BLDPBD161281-10G
2-Chlor-6-hydroxybenzonitril 97%
2-Chlor-6-hydroxybenzonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₄ClNO
Molekulargewicht: 153,5658 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01646165
CAS-Nummer: 89999-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR