3-Fluor-2-nitrobenzoesäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD161300-100GEA 1036 EUR
BLDPBD161300-100G BLDPBD161300-10G BLDPBD161300-250MG BLDPBD161300-100MG BLDPBD161300-25G BLDPBD161300-1G BLDPBD161300-5G
3-Fluor-2-nitrobenzoesäure 98%
3-Fluoro-2-nitrobenzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₄FNO₄
Molekulargewicht: 185,11 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07368342
CAS-Nummer: 1000339-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR