4-Bromo-3-methylbenzyl bromide 95%
|
|
BLDPBD161762-1GEA
55
EUR
BLDPBD161762-1G
BLDPBD161762-25G
BLDPBD161762-5G
BLDPBD161762-250MG
BLDPBD161762-100MG
4-Bromo-3-methylbenzyl bromide 95%
4-Bromo-3-methylbenzyl bromide
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₈H₈Br₂ Molekulargewicht: 263,957 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09909629 CAS-Nummer: 27561-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...