4-Methoxycarbonyl-3-nitrophenylboronic acid 98%

BLDPBD161836-10GEA 519 EUR
BLDPBD161836-10G BLDPBD161836-1G BLDPBD161836-250MG BLDPBD161836-5G
4-Methoxycarbonyl-3-nitrophenylboronic acid 98%
4-Methoxycarbonyl-3-nitrophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₈BNO₆
Molekulargewicht: 224,963 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD12032027
CAS-Nummer: 85107-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR